Find methods for your needs
AN73852: Determination of Trace Anions in High Purity Waters using a Large-Volume Direct Injection
Description
This work describes the use of Dionex IonPac AS28-Fast-4μm column that was designed specifically for the trace analysis of inorganic anions and monovalent organic acids. Improved peak efficiencies in combination with the smaller column inner diameter result in lower detection limits compared to the Dionex IonPac AS15-5μm column. The analytes are detected by suppressed conductivity with a Dionex ADRS™ 600 suppressor. This application note expands on work presented in Application Update 142, “Improved determination of trace anions in high purity waters by high-volume direct injection| Market: | Electronics & Semiconductors |
| Keywords: | Dionex ICS-5000+ System, ion chromatography, IC, Chloride, Sulfate, semiconductor, HPW, Dionex EGC 500 KOH eluent generator cartridge, Dionex IonPac AS28-Fast-4μm column, Dionex ICS-6000 system, AN 73852, TN 73892, TN73892 |
| Matrix: | High purity water |
| Author: | Manali Aggrawal and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.