Find methods for your needs
AN73827: Determination of Nitrite and Nitrate in Sugar using Ion Chromatography Be the first to rate this application
Description
Natural sugar products are material used for food and animal feed. Nitrite and nitrate are undesirable substances in sugar products. Here, an IC method was developed and validated using a 2 mm version of the Dionex IonPac AS11-HC-4μm column for the determination of nitrite and nitrate in sugar samples. The method is sensitive and accurate. It can be used to determine nitrite and nitrate in sugar products for regulatory monitoring.
Market: Food and Beverage
Keywords: Animal feed, food, beet sugar, RFIC System, suppressed conductivity detection, Dionex IonPac AS11-HC-4μm column, AN 73827, molasses
Matrix: Sugar
Author: Hua Yang and Jeffrey Rohrer
Affiliation: Thermo Fisher Scientific, Sunnyvale, CA USA
Uploaded on 10/19/2020.