Find methods for your needs
AB73728: Low-Cost Determination of Inorganic Cations and Ammonium in Environmental Waters
Description
This work demonstrates the determination of cations and ammonium in environmental waters by IC using a Thermo Scientific™ Dionex™ IonPac™ CS16 column set on a Thermo Scientific™ Dionex™ EASION chromatography system in DCR (Displacement Chemical Regeneration) mode. The Dionex EASION IC system is an integrated, single-channel low-cost IC system designed for isocratic applications with suppressed conductivity detection. Coupled to the Thermo Scientific™ Dionex™ AS-DV Autosampler, it provides a low-cost choice for routine water analysis.| Market: | Environmental |
| Keywords: | Dionex IonPac CS16 Column, wastewater, Drinking water, AS-DV autosampler, Easion |
| Matrix: | Water |
| Author: | Hua Yang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |