Find methods for your needs
AB73726: Fast and Low-Cost Determination of Anions in Municipal Drinking Water
Description
This work demonstrates the determination of inorganic anions in drinking water by IC using a Thermo Scientific™ Dionex™ IonPac™ AS22-Fast-4μm column set on a Thermo Scientific™ Dionex™ EASION Chromatography system in DCR (Displacement Chemical Regeneration) mode. The Dionex EASION IC system is an integrated, single-channel low-cost IC system designed for isocratic applications with suppressed conductivity detection. Coupled to the Thermo Scientific™ Dionex™ AS-DV Autosampler, it provides a fast and low-cost instrument setup for routine determination of inorganic anions in drinking water.| Market: | Environmental |
| Keywords: | Drinking water, AS-DV autosampler, U.S. EPA Method 300.1, Easion, Dionex IonPac AS22-Fast-4μm column |
| Matrix: | Water |
| Author: | Hua Yang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |