Find methods for your needs
A fast, cost-effective HS-GC-FID method for the analysis of ethylene oxide in surgical-style face masks
Description
A fast, cost-effective analytical method suitable for the analysis of residual ethylene oxide and 2-chloroethanol in face masks using static headspace (HS) gas chromatography (GC) with flame ionization detection (FID).| Market: | Other |
| Keywords: | FID, GC, Headspace, Flame Ionization Detection, Gas Chromatography, valve and loop, TriPlus 500 HS, carcinogenic, ethylene oxide, face masks, GB 19083-2010, GBT 16886.7-2015 |
| Matrix: | Face masks |
| Author: | David Lee; Cristian Cojocariu |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.