Find methods for your needs
Charge variant analysis of Ranibizumab sample using volatile buffer and strong cation exchange column.
Description
The Thermo Scientific Dionex UltiMate 3000 LC Bio-LC system is applied for the Ion Exchange analysis of ranibizumab sample. The separation was performed on a Thermo Scientific MAbPac SCX-10 with a UV detection at 280 nm.| Market: | BioPharma |
| Keywords: | HPLC, charge variant analysis, MAbPac SCX 10, MS compatible, Ranibizumab, CVA MS |
| Matrix: | N.A |
| Author: | Ankitkumar Patel |
| Affiliation: | Thermo Fisher Scientific, CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.