Description
Demonstration of an analytical method that meets the requirements outlined in U.S. EPA Method 8260D utilizing U.S. EPA Methods 5030 and 5035 preparation methods for the quantitation of purgeable organic compounds (POCs) in wastewater and soil. Method linearity, method detection limit (MDL), and Initial Demonstration of Capability (IDC) were assessed to evaluate method performance.| Market: | Environmental |
| Keywords: | EPA, GRO, trace analysis, Gas Chromatography, selected ion monitoring, single quadrupole mass spectrometry, environmental sample analysis, THM, VOCs, analytical testing laboratories, environmental laboratories, P&T |
| Matrix: | Soil, Wastewater |
| Author: | Adam Ladak, Terry Jeffers, Dwain Cardona, David Lee, and Amy Nutter |
| Affiliation: | Thermo Fisher Scientific, Teledyne Tekmar |
For Research Use Only. Not for use in diagnostic procedures.