Find methods for your needs
Analysis of VOCs in wastewater using P&T-single quadrupole GC-MS
Description
Show proof of principle for the major method challenges of U.S. EPA Method 624 for the quantitation of volatile organic compounds (VOCs) in wastewater, using the Teledyne Tekmar Atomx XYZ purge and trap (P&T) system along with a Thermo Scientific ISQ7000 MS system coupled with a Thermo Scientific TRACE1310 GC and Thermo Scientific Chromeleon Chromatography Data System (CDS). Linearity, method detection limit (MDL), precision, accuracy, and Initial Demonstration of Capability (IDC) were assessed to evaluate method performance.| Market: | Environmental |
| Keywords: | environmental, EPA, trace analysis, wastewater, Gas Chromatography, selected ion monitoring, single quadrupole mass spectrometry, environmental sample analysis, purge and trap, VOCs, contract testing labs, environmental testing lab |
| Matrix: | Wastewater |
| Author: | Adam Ladak, Terry Jeffers, David Lee, and Amy Nutter |
| Affiliation: | Thermo Fisher Scientific, Teledyne Tekmar |
For Research Use Only. Not for use in diagnostic procedures.