Find methods for your needs
Related substances analysis of sumatriptan succinate using a mixed-mode HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of the related substances of sumatriptan succinate. The analysis was performed on a Thermo Scientific Acclaim Mixed-Mode WCX-1 HPLC column. The separation results and peak asymmetry were satisfactory.| Market: | Pharma |
| Keywords: | HPLC, UV detection, sumatriptan, mixed-mode column |
| Matrix: | N/A |
| Author: | Krishna rao dara |
| Affiliation: | thermo Scientific, CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.