Find methods for your needs
Related substances analysis of metformin using a HILIC HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of the related substances of metformin. The analysis was performed on a Thermo Scientific Syncronis HILIC column. All four known impurities were well resolved from metformin.| Market: | Pharma |
| Keywords: | HILIC, HPLC, Metformin, UV detection, biguanides |
| Matrix: | N/A |
| Author: | Krishna rao dara |
| Affiliation: | thermo Scientific, CoE Lab, Ahmedabad, India |