Find methods for your needs
Analysis of trace antimony in water - Complying with Chinese standard methods for atomic absorption spectroscopy - HJ 1046-2019 and HJ 1047-2019
Description
To demonstrate the applicability of the Thermo Scientific™ iCE™ 3300 Flame AAS and Thermo Scientific™ iCE™ 3400 Graphite Furnace AAS for determination of antimony in diverse water samples based on Chinese environmental standard methods (HJ 1046-2019, HJ 1047-2019) with different application focus.Market: | Environmental; Water Analysis |
Keywords: | surface water, Antimony analysis, flame AAS, graphite furnace AAS, hot plate digestion, industrial wastewater |
Matrix: | Industrial wastewater samples, River water samples |
Author: | Jianfeng Cui, Tomoko Vincent, and Bhagyesh Surekar |