Find methods for your needs
AN73745: Determination of Sodium, Potassium, and Calcium in Rice and Wheat Flours Using Ion Chromatography
Description
Here, we employed IC to measure calcium, potassium,and sodium in rice and wheat flours. The method uses a Thermo Scientific™ Dionex™ IonPac™ CS16 column, an electrolytically generated methanesulfonic acid (MSA) eluent, and suppressed conductivity detection on a Reagent-Free™ Ion Chromatography (RFIC™) system.| Market: | Food and Beverage |
| Keywords: | Dionex IonPac CS16 Column, suppressed conductivity detection, RFIC system |
| Matrix: | Rice Flour, Wheat flour |
| Author: | Beibei Huang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.