Find methods for your needs
Robust analysis of REE in electronic waste - High sample throughput with analysis times of less than 2 minutes per sample with the iCAP PRO Series ICP-OES Be the first to rate this application
Description
This application note describes the analysis of rare earth elements (REE), precious metals and toxic elements in electronic waste using the iCAP PRO XP Duo ICP-OES instrument. The screens, circuit boards and speaker magnets of two mobile phones were analyzed and different concentrations of REEs and other metals were found in them. Fast analysis, high sensitivity in complicated matrix, large linear dynamic range, and excellent system robustness was observed over the course of the analyses making the iCAP PRO ICP-OES the ideal choice for such applications in routine laboratories.
Market: Environmental; Metals & Mining; Other
Keywords: recycling, REE, Environment, high matrix, ICP-OES, WEEE, robust, analytical testing, Electronic waste, mobile phones, rare earth elements
Matrix: Electronic waste, mobile phone thin-film transistor screen, printed circuit boards, speaker magnets
Author: Sukanya Sengupta, Cristian Cojocariu, Bhagyesh Surekar
Uploaded on 8/3/2020.