Find methods for your needs
AN73608: Determination of Four Polar Pesticides in Drinking Water by IC-MS
Description
This study describes the simultaneous direct determination of endothall, glyphosate, AMPA, and glufosinate in drinking water by IC-MS. The four pesticides can be determined sensitively and accurately using a Dionex IonPac AS19-4μm column and an ISQ EC single quadrupole mass spectrometer.| Market: | Environmental |
| Keywords: | AMPA, Glyphosate, endothall, Dionex IonPac AS19-4μm column, Single quadrupole mass spectrometer, Dionex Integrion HPIC, ISQ EC, TN72611, glufosinate, Dionex ADRS 600 suppressor, AN 73608, TN 72611 |
| Matrix: | Water |
| Author: | Jingli Hu and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.