Find methods for your needs
Analysis of VOCs in automotive trimcomponents using TD-GC-MS
Description
To demonstrate a simple and effective analytical solution for the direct analysis of volatile organic compounds (VOC) and semi volatile condensable compounds (FOG) emitted from car interior trim material referencing base parameters in the VDA 278 method, utilizing the thermal desorption sampling technique coupled to GC-MS and Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS) software for data acquisition, processing, and intelligent reporting against hazardous chemical lists.| Market: | Chemical; Material Science |
| Keywords: | GC-MS, mass spectrometry, Gas Chromatography, single quadrupole mass spectrometry, ISQ 7000, volatile organic compounds, Automotive industry, automotive interior materials, FOG, GADSL, Global Automotive Declarable Substance List, hazardous chemicals, REACH, semi volatile organic substances, SVOC, TD, TD100-xr, thermal desorption analysis, vehicle interior air quality, VIAQ, VOC, volatile condensible compounds |
| Matrix: | automotive interior materials |
| Author: | Klaus Schrickel (1) and Jane Cooper (2) |
| Affiliation: | (1) Thermo Fisher Scientific, Germany; (2) Thermo Fisher Scientific, Runcorn, UK |
For Research Use Only. Not for use in diagnostic procedures.