Find methods for your needs
AN73607: Determination of Inorganic Anions in Drinking Water, Wastewater, and High Ionic Strength Water
Description
This application note (AN) demonstrates the determination of inorganic anions in water samples using a Thermo Scientific™ Dionex™ IonPac™ AS29-Fast-4μm anion-exchange column. The AN also show the benefits of this column compared to a Thermo Scientific™ Dionex™ IonPac™ AS4A-SC column, the column originally specified in U.S. EPA Method 300.0 Part A. It was found that Dionex IonPac AS29-Fast-4μm column has improved selectivity and higher capacity, making it a better choice for compliance monitoring of inorganic anions, especially for high-ionic strength water and other challenging samples.| Market: | Environmental |
| Keywords: | suppressed conductivity detection, RFIC System, Dionex IonPac AS29-Fast-4μm column, Safe Drinking Water Act (SDWA), Clean Water Act (CWA), U.S. EPA Method 300.1, AN 73607, EPM, K2CO3 |
| Matrix: | Water |
| Author: | Hua Yang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.