Find methods for your needs
Rapid determination of phthalates in polymers, maximizing sample throughput to <10 min per sample using pyrolyzer/thermal desorption GC-MS
Description
Establish a simple and highly automated analytical method to detect and quantify phthalates in polymers with a view to providing an improved rapid screening method.| Market: | Chemical; Pharma; Food and Beverage; Electronics & Semiconductors; Material Science |
| Keywords: | GC-MS, Polymer, Phthalate, mass spectrometry, Gas Chromatography, single quadrupole mass spectrometry, ISQ 7000, thermal desorption, pyrolyzer |
| Matrix: | Plastics |
| Author: | Yan Weixian (1) and Jane Cooper (2) |
| Affiliation: | (1) Thermo Fisher Scientific, China; (2) Thermo Fisher Scientific, Runcorn, UK |
For Research Use Only. Not for use in diagnostic procedures.