Description
The goal of the methodology described here was to demonstrate the suitability of a simplified, cryogen-free method for the analysis of VOCs in exhaust gases, using thermal desorption (TD) coupled to dual-column gas chromatography-flame ionization (GC–FID) detection.Market: | Environmental |
Keywords: | FID, GC, Gas Chromatography, air toxics, CIA Advantage-xr canister autosampler, cryogen-free monitoring, Flame ionization detector, Kori-xr water removal device, PAMS, photo assessment monitoring scheme, VOCs, air pollution, Deans Switch, thermal desorption, volatile organic compounds |
Matrix: | Exhaust gases |
Author: | David Lee; Terry Jeffers; Cristian Cojocariu |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.