Find methods for your needs
Enantiomeric purity (%) of l-Ephedrine sulphate by HPLC-CSP
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the enatioselective separation of ephedrine sulphate. The analysis was performed on a Thermo Scientific Hypersil Chiral -OT column. The results obtained for resolution between the peaks due to l-ephedrine and d-ephedrine was above 3.| Market: | Pharma |
| Keywords: | HPLC, UV detection, Chiral, CSP, Ephedrine |
| Matrix: | N/A |
| Author: | Krishna rao dara |
| Affiliation: | thermo Scientific, CoE Lab, Ahmedabad, India |