Find methods for your needs
Enantiomeric purity (%) of l-Ephedrine sulphate by HPLC-CSP Be the first to rate this application
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the enatioselective separation of ephedrine sulphate. The analysis was performed on a Thermo Scientific Hypersil Chiral -OT column. The results obtained for resolution between the peaks due to l-ephedrine and d-ephedrine was above 3.
Market: Pharma
Keywords: HPLC, UV detection, Chiral, CSP, Ephedrine
Matrix: N/A
Author: Krishna rao dara
Affiliation: thermo Scientific, CoE Lab, Ahmedabad, India
Uploaded on 6/1/2020.