Find methods for your needs
Fast and Reproducible peptide mapping of an Pertuzumab (IgG1) by using a SMART digest Kit and fully porous 120 A C18 column.
Description
The Thermo Scientific Dionex Ultimate 3000 system is applied for the peptide mapping analysis of an Pertuzumab (IgG1). The separation was performed on a Thermo Scientific Acclaim RSLC 120 C18 column by using a Thermo Scientific SMART digest kit at UV detection of 214 nm. The SMART Digest kit provides significant improvements in reproducibility, which results in fewer sample failures, higher throughput and the ability to more easily interrogate data..| Market: | BioPharma |
| Keywords: | Peptide mapping, UV detection, UHPLC, Trypsin digestion, SMART Digest kit, IgG1, Pertuzumab |
| Matrix: | NA |
| Author: | Ankitkumar Patel |
| Affiliation: | Thermo Fisher Scientific, CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.