Find methods for your needs
AU73278: Improved Ion Chromatography Column for Separation of Ethylenediamine Carbamate and Fluoride, and Carbonate and Sulfate in Drinking Water
Description
This study demonstrates the ability of the Thermo Scientific™ Dionex™ IonPac™ AS30 column to separate ethylenediamine (EDA) carbamate and fluoride as well as carbonate and sulfate in order to determine trace concentrations of oxyhalides and bromide in drinking water containing the EDA preservative.| Market: | Environmental |
| Keywords: | Bromate, Dichloroacetate, Disinfection Byproducts, EDA, Oxyhalides, suppressed conductivity detection, Dionex IonPac AS30 column, AU 73278, EPA Method 300.1 Part B |
| Matrix: | Drinking water |
| Author: | Beibei Huang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.