Find methods for your needs
Confident quantitation for every user: immunosuppressants in human blood by liquid chromatography-tandem mass spectrometry for clinical research
Description
Implementation of an analytical method for the quantification of four immunosuppressants (tacrolimus, sirolimus, everolimus, and cyclosporine A) in human blood on a Thermo Scientific™ TSQ Quantis™ triple quadrupole mass spectrometer| Market: | Clinical Research |
| Keywords: | Blood, multi-channeling, tacrolimus, sirolimus, everolimus, mass spectrometry, TSQ Quantis, Immunosuppressants, offline sample preparation, cyclosporine A |
| Matrix: | Human blood |
| Author: | Claudio De Nardi,1 Paolo Brambilla,2 Maura Brambilla,2 Chiara Fania3 |
| Affiliation: | 1 Thermo Fisher Scientific GmbH, Dreieich, Germany; 2 Ospedale di Desio, Desio, Italy; 3 Università degli Studi MilanoBicocca, Milano, Italy |
For Research Use Only. Not for use in diagnostic procedures.