Find methods for your needs
New Double Barrel ESI Source and Novel Tandem NanoLC-MS for 24/7 Proteome Profiling with near 100% MS Utilization
Description
The developed tandem nanoLC-MS platform which allows routine 24/7 operation with close to 100% MS utilization comprises: (i) tandem Thermo Scientific™ UltiMate™ 3000 RSLCnano system; (ii) Double Barrel Column Oven (Sonation GmbH) installed onto the Thermo Scientific™ Nanospray Flex Ion Source; (iii) Thermo Scientific™ Q Exactive™ HF-X Hybrid Quadrupole-Orbitrap™ Mass Spectrometer; (iv) Optimized intelligent method script for automated switching between columns during the method execution| Market: | Proteomics |
| Keywords: | Proteomics, LCMS, Orbitrap, productivity, Tandem, nanoLCMS, Utilization, Dual Source |
| Matrix: | HeLa, Plasma, Serum, Tissue |
| Author: | Runsheng Zheng |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.