Find methods for your needs
CAN73481: AOF by Combustion IC – Non-Targeted Complemental Determination of PFAS in Aqueous Samples
Description
PFAS are human-made chemicals, which have been manufactured since the 1940s. The most well-known PFAS compounds, perfluorooctanoic acid (PFOA) and perfluorooctanesulfonic acid (PFOS), have been extensively studied for their chemical properties and toxicological effects. PFAS are used in a wide variety of consumer products, such as nonstick cookware, food containers, stain- and water-repellent fabrics, polishes, waxes, paints, and cleaning products. This work focuses determing PFAS, and the adsorbable organically bound fluorine (AOF) by Combustion Ion Chromatography| Market: | Environmental |
| Keywords: | RFIC, ion chromatography, IonPac AS20 column, PFOA, PFAS, AN 72333, suppressed conductivity, AN72333, AOX, CAN 73481, gradient elution, perfluorinated organic substances, polyfluorinated organic substances, polyfluoroalkyl substances, perfluoroalkyl, TN000767 substances |
| Matrix: | Drinking water, Wastewater |
| Author: | Eleonora von Abercron, Udo Neist, Inge Klocke, Sebastian Georgii, Hubertus Brunn, Detlef Jensen (1) |
| Affiliation: | Hessian State Laboratory, Wiesbaden, Germany; (1)- Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.