Find methods for your needs
AN73390: Determination of Haloacetic Acids, Bromate and Dalapon in Drinking Water using Ion Chromatography Coupled to High-Resolution Accurate-Mass (IC-HRAM) Mass Spectrometry
Description
This application note demonstrates the capability and performance of a Thermo Scientific™ Q Exactive™ HF hybrid quadrupole-Orbitrap mass spectrometer-based IC-HRAM MS method to identify and quantify nine haloacetic acids, bromate, and dalapon in drinking water.| Market: | Environmental |
| Keywords: | Disinfection Byproducts, HAAs, Dionex ICS-6000, Dionex IonPac AS31 column, Q Exactive HF Hybrid Quadrupole-orbitrap |
| Matrix: | Drinking water |
| Author: | Beibei Huang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.