Find methods for your needs
Ion exchange analysis of teriperatide using polymeric non-porous strong cation exchange column.
Description
The Thermo Scientific Dionex UltiMate 3000 LC Bio-LC system is applied for the Ion Exchange analysis of teriperatide. The separation was performed on a Thermo Scientific MAbPac SCX-10 and fluorescence detector.| Market: | BioPharma |
| Keywords: | HPLC, MAbPac SCX 10, ion exchange analysis, PTH, teriperatide |
| Matrix: | NA |
| Author: | Ankitkumar Patel |
| Affiliation: | thermo Fisher Scientific, CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.