Find methods for your needs
Fast analysis of contaminants in soil using a Thermo Scientific Hypersil GOLD CN HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of contaminants in soil. The separation is performed on a Thermo Scientific Hypersil GOLD CN HPLC column, with UV detection at 254 nm.| Market: | Environmental |
| Keywords: | Benzyl alcohol, Contaminants, Ethyl benzene, HPLC, Hypersil GOLD CN, o-Cresol, p-Hydroxybenzaldehyde, soil, Toluene, UV detection |
| Matrix: | Soil |
For Research Use Only. Not for use in diagnostic procedures.