Find methods for your needs
AN73389: Determination of Sodium, Potassium, and Calcium in Dietary Vitamins and Sauerkraut
Description
This application used the Dionex IonPac CS16 column with a 30 mM MSA eluent at 40 °C and suppressed conductivity detection to determine inorganic cations in multivitamin and sauerkraut samples at concentrations ranging from 1.98 to 200 mg/L. The high capacity of the column allows loading of high concentrations of sodium, potassium, and calcium without affecting/losing resolution.| Market: | Food and Beverage |
| Keywords: | Dionex IonPac CS16 Column, Potassium, Calcium, Sodium, Suppressed Conductivity, Dionex ICS-6000, multivitamin tablets, sauerkraut, ion chromatography, AN 73389 |
| Matrix: | multivitamins, sauerkraut |
| Author: | Manali Aggrawal and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.