Find methods for your needs
Determination of Trace Anions using Thermo Scientific Dionex AutoPrep on the Thermo Scientific Dionex ICS-6000 HPIC Ion Chromatography System: Large Loop
Description
Demonstrates the configuration and use of Dionex AutoPrep configured on the Dionex ICS-6000 system with AS-HV autosampler and auxiliary pump. The system is controlled by Thermo Scientific Chromeleon 7 version 3| Market: | Electronics & Semiconductors |
| Keywords: | AS-HV autosampler, large volume injection, fluoride, Nitrate, RFIC, Nitrite, Chloride, phosphate, IonPac AS17-C, Acetate, Sulfate, ultra pure water, Reagent-Free IC, Bromide, AutoPrep, Dionex IonPac AS17-C, large loop, Large Loop Small Loop, small loop |
| Matrix: | Ultra pure water |
| Author: | Terri Christison, Jay Lorch, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
| Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.