Find methods for your needs
AN73346: Determination of Per- and Polyfluorinated Alkyl Substances (PFAS) in Drinking Water using Automated Solid-Phase Extraction and LC-MS/MS
Description
This application note presents an efficient and reliable solid-phase extraction method with the Thermo Scientific™ Dionex™ AutoTrace™ 280 instrument for the determination of perfluorinated and polyfluorinated compounds in drinking water per U.S. EPA Method 537.1. The manual solid phase extraction is replaced with a reproducible, reliable and automated AutoTrace 280 SPE system.
Market: Environmental
Keywords: UHPLC, PFOA, PFOS, AutoTrace 280, perfluorinated alkyl substances, PFAS, TSQ Fortis triple quadrupole mass spectrometer, Accucore RP-MS column, U.S. EPA Method 537.1, PFHxS, Vanquish Flex Duo, PFTA, PFTrDA, NMeFOSAA, PFUnA, 9Cl-PF3ONS, PFDoAPFBS, PFDA, PFNA, PFHpA, PFDoA
Matrix: Water
Author: Changling Qiu, Xin Zhang, Rahmat Ullah, Wei Chen, Yan Liu
Affiliation: Thermo Fisher Scientific
Uploaded on 2/7/2020.