Find methods for your needs
Fast and Reproducible peptide mapping of an peptibody (romiplostim) by using a SMART digest Kit and solid core C18 column
Description
The Thermo Scientific Dionex Ultimate 3000 system is applied for the peptide mapping analysis of an peptibody (Romiplostim). The separation was performed on a Thermo Scientific Vanquish C18+ column by using a Thermo Scientific SMART digest kit at UV detection of 210 nm. The SMART Digest kit provides significant improvements in reproducibility, which results in fewer sample failures, higher throughput and the ability to more easily interrogate data.| Market: | BioPharma |
| Keywords: | Peptide mapping, UV detection, UHPLC, Trypsin digestion, SMART Digest kit, Romiplostim, Peptibody |
| Matrix: | N/A |
| Author: | Ankitkumar Patel |
| Affiliation: | thermo Fisher Scientific, CoE Lab, Ahmedabad, India |