Find methods for your needs
AN73342: Determination of Low-Level Haloacetic Acids, Bromate, and Dalapon in Drinking Water using IC-MS
Description
This AN evaluated an IC-MS method for the determination of all nine haloacetic acids, bromate, and dalapon in drinking water using a Dionex ICS-6000 HPIC system couple with an ISQ EC single quadrupole mass spectrometer. Dionex IonPac AS31 column was used. It found that the IC-MS method is fast (40 min vs. 60 min in Method 557) and linear. It is sensitive (<1 μg/L), accurate and precise for the determination of U.S. EPA regulated HAA5, bromate, and dalapon. Critical IC system conditions important for successfully running this application are included.Market: | Environmental |
Keywords: | Disinfection Byproducts, Monochloroacetic acid (MCAA), HAA5, HAAs, ISQ EC, eWorkflow, Dionex IonPac AS31 column, Single quadrupole MS, bromochloroacetic acid (BCAA), dibromoacetic acid (DBAA), dichloroacetic acid (DCAA), monobromoacetic acid (MBAA), trichloroacetic acid (TCAA), bromodichloroacetic acid (BDCAA), chlorodibromoacetic acid (CDBAA), tribromoacetic acid (TBAA), U.S. EPA Method 557 |
Matrix: | Water |
Author: | Hua Yang and Jeffrey Rohrer |
Affiliation: | Thermo Fisher Scientific |