Find methods for your needs
AN73341: Determination of Sugars in Dairy Products using HPAE-PAD Be the first to rate this application
Description
This application notes demonstrates that ISO/DIS 22184 for sugars (galactose, glucose, fructose, sucrose, lactose and maltose) determination in dairy products can be executed with a Dionex CarboPac PA1-2mm column using a Dionex HPIC system.
Market: Food and Beverage
Keywords: carbohydrate, Electrochemical detection, Milk, Infant Formula, Cheese, Dionex ICS 5000+ HPIC system, Dionex ICS-6000 HPIC system, TN71, Dionex CarboPac PA1-2mm column, ISO, whey powder, AU 73341, TN 71, TN 20, TN 70671, TN20, TN70671
Matrix: cheese, Infant formula, Milk, whey powder
Author: Jingli Hu and Jeffrey Rohrer
Affiliation: Thermo Fisher
Uploaded on 1/31/2020.
For Research Use Only. Not for use in diagnostic procedures.