Find methods for your needs
Global round robin test of thiopental EP method performance on identical HPLC systems Be the first to rate this application
Description
The presented results show that Thermo Scientific™ Vanquish™ Core HPLC systems have excellent system-to system reproducibility when the sample preparation, eluent preparation, and LC column are identical. Users can rely on the fact that the performance of multiple Vanquish Core HPLC systems will be highly predictable and robust for routine methods in quality control labs when other variables are controlled.
Market: Pharma
Keywords: reproducibility, Precision, European Pharmacopoeia, system suitability test, thiopental, UV-Vis, variability, small molecule pharmaceutical
Matrix: Acetonitrile, Phosphoric acid
Author: Sylvia Grosse, Katherine Lovejoy, Frank Steiner
Affiliation: Thermo Fisher Scientific, Germering, Germany
Uploaded on 1/31/2020.
For Research Use Only. Not for use in diagnostic procedures.