Find methods for your needs
TN73348: Carbohydrate Determinations by HPAE-PAD using a PdH Reference Electrode
Description
We recently introduced a PdH RE for HPAE-PAD carbohydrate determinations. In this work, we demonstrate two popular carbohydrate applications (honey sugars and glycoprotein monosaccharides) using a PdH RE and compare the results to using an Ag/AgCl RE. Our preliminary data indicate that the PdH RE provides comparable results to the standard Ag/AgCl RE for these two applications that use hydroxide eluents.| Market: | BioPharma; Food and Beverage |
| Keywords: | Electrochemical detection, fucose, Glucose, glycoprotein, Monosaccharides, Mannose, Maltose, Sucrose, trehalose, fructose, galactosamine, glucosamine, ICS-6000, TN71, Dionex CarboPac PA20-4μm colum, Dionex CarboPac PA210-4μm column, honey sugars, isomaltose, turanose, 1-kestose, melezitose, erlose, TN 73348, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Bovine serum Fetuin, Honey |
| Author: | Manali Aggrawal; Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.