Find methods for your needs
AN71993: Profiling Galactosyloligosaccharide-Containing Samples by High-Performance Anion-Exchange Chromatography with Pulsed Amperometric Detection (HPAE-PAD)
Description
GOS occur naturally in the milk of many animals including humans, cows, and wallabies. GOS are primarily composed of galactose and often terminate with a glucose residue at the reducing end. These carbohydrates are enzymatically produced by transgalactosylation reactions of lactose catalyzed by β-galactosidases and give rise to galactose oligomers. Separation of individual oligosaccharides in GOS was achieved on a Dionex™ CarboPac™ PA200 column. Oligosaccharide detection was by PAD with a gold working electrode, and therefore no sample derivation was required. This replaced AN1151.| Market: | Food and Beverage |
| Keywords: | Carbohydrates, GOS, oligosaccharide, Infant Formula, prebiotics, probiotics, TN21, TN71, Dionex CarboPac PA200, disposable electrode, AN 71993, AN 1151, AN1151, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Infant formula |
| Author: | Manali Aggrawal; Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.