Find methods for your needs
AN73051: Determination of Bromate in Flour and Bread by IC-MS
Description
Potassium bromate is a food additive used as “flour improver” in the baking industry. Bromate is considered a carcinogenic and nephrotoxic substance. We developed a method for a selective and sensitive determination of bromate in flour that uses IC coupled with MS. A recently introduced high-capacity anion-exchange column was used to separate bromate from matrix anions. Six commercial flour and flour products including homemade bread baked using flour containing potassium bromate, were analyzed. LOD and LOQ of bromate in the prepared solution were 0.10 µg/L and 0.34 µg/L, respectively.Market: | Food and Beverage |
Keywords: | Bromate, IC-MS, mass spectrometry, oxyhalide, bread, IonPac AS31 column, ISQ EC single quadrupole mass spectrometer, flour, AN 73051 |
Matrix: | Flour and flour prodcuts |
Author: | Manali Aggrawal; Jeff Rohrer |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.