Find methods for your needs
Separation of charge variants of romiplostim by Ion Exchange chromatography using strong cation exchange column
Description
The Thermo Scientific Dionex UltiMate 3000 LC Bio-LC system is applied for the Ion Exchange analysis of romiplostim i.e peptibody. The separation was performed on a Thermo Scientific MAbPac SCX-10 with a UV detection at 210 nm.| Market: | BioPharma |
| Keywords: | HPLC, charge variant analysis, Romiplostim, MAbPac SCX 10, Peptibody |
| Matrix: | NA |
| Author: | Ankitkumar Patel |
| Affiliation: | thermo Fisher Scientific, CoE Lab, Ahmedabad, India |