Find methods for your needs
Oxidised impurity analysis of Denosumab using HIC technique.
Description
The Thermo Scientific Dionex ultimate 3000 Bio compatible LC system is applied for the oxidised impurities analysis of Denosumab.The analysis was performed on a Thermo Scientific MAbPac HIC 20 column. Please Note: Blank normalized chromatogram is shown in representative profile.| Market: | BioPharma |
| Keywords: | HPLC, Denosumab, HIC, IgG2, MAbPac HIC 20 |
| Matrix: | NA |
| Author: | Ankitkumar Patel |
| Affiliation: | thermo scientific, CoE Lab, Ahmedabad, India |