Find methods for your needs
AN73280: Determination of Fluorine and Chlorine in Iron Ore using Combustion Ion Chromatography
Description
This application note developed a CIC method for determining fluorine and chlorine in iron ore samples. Key performance parameters were evaluated including separation, linearity, limits of detection, and precision. Fluorine and chlorine concentrations were determined in six iron ore samples.| Market: | Metals & Mining |
| Keywords: | CIC, AQF-2100H, suppressed conductivity detection, Dionex Integrion HPIC System, ADRS 600 Suppressor, Dionex IonPac AS18-Fast-4μm column, mining, AN 73280, TN000767, AN73280 |
| Matrix: | Iron Ore |
| Author: | Jingli Hu and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.