Find methods for your needs
Online 2D-nanoLC hyphenated with high-resolution accurate-mass Orbitrap mass spectrometry for comprehensive and robust proteome profiling Be the first to rate this application
Description
In this technical note, we developed and elucidated the benefits of an online 2D nanoLC-MS platform for proteomics. We present a simple-to-use online low-flow high-pH RP x low-pH RP separation platform (termed online 2D-nanoLC) for deep proteome profiling using a Thermo Scientific™ UltiMate™ 3000 RSLCnano system coupled to a Thermo Scientific™ Orbitrap Exploris™ 480 mass spectrometer. This automated approach not only reduces sample complexity but also boosts peptide/protein identifications and quantification without extra sample handling steps.
Market: Proteomics
Keywords: Proteomics, Q Exactive HF-X, nanoLCMS, UltiMate 3000 RSLCnano, Orbitrap Exploris 480, Deep Proteome Profiling, online 2D-LC
Matrix: HeLa Cell Lysate Protein Digest, Serum Protein Digest
Author: Runsheng Zheng, Alexander Boychenko, Tabiwang N. Arrey, Christopher Pynn, Wim Decrop, Peter Jehle
Affiliation: Thermo Fisher Scientific, Germering, Germany; Thermo Fisher Scientific, Bremen, Germany
Status: Proof of Concept
Uploaded on 11/4/2019.
For Research Use Only. Not for use in diagnostic procedures.