Find methods for your needs
Quantitative determination of seven synthetic cathinones (stimulants) from stabilized human urine by UHPLC-MS/MS for forensic toxicology
Description
Quantitative analytical method for the determination of seven synthetic cathinones from stabilized human urine. The Thermo Scientific™ Syncronis™ HILIC 1.7 μm column is used in combination with the Thermo Scientific™ Vanquish™ Horizon UHPLC system and Thermo Scientific™ TSQ Quantiva™ triple-stage quadrupole MS/MS system. Thermo Scientific™ SOLA™ SCX solid phase extraction technology is used for sample clean-up prior to analysis.| Market: | Forensics; Toxicology |
| Keywords: | Bath salts, HILIC, SOLA SCX, Syncronis, Urine, TSQ Quantiva, Cathinones, bioanalysis, Vanquish Horizon, triple quad, khat, legal highs, plant food |
| Matrix: | Urine |
| Author: | Matt Franklin and Kean Woodmansey |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.