Find methods for your needs
Separation of related impurities of Romiplostim by Reverse Phase chromatography using a wide pore phenyl column.
Description
The Thermo Scientific Dionex UltiMate 3000 LC Bio-LC system equipped with a column pre-heater is applied for the Reverse phase analysis of Romiplostim. The separation was performed on a Thermo Scientific MAbPac RP column with UV detection at 280 nm.| Market: | BioPharma |
| Keywords: | HPLC, MAbPac RP, Oxidation, Romiplostim |
| Matrix: | N.A |
| Author: | Ankitkumar Patel |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |