Find methods for your needs
Assay and organic impurities analysis of Buprenorphine and Naloxone sublingual tablets using a Phenyl HPLC column as per USP 42 monograph
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for assay and organic impurities analysis of buprenorphine and naloxone sublingual tablets. The analysis was performed on a Thermo Scientific Syncronis Phenyl HPLC column using the method described in the USP 42. The observed results met the system suitability criteria for both assay and organic impurities test parameters.| Market: | Pharma |
| Keywords: | HPLC, UV detection, Buprenorphine, Naloxone, Opiods |
| Matrix: | N.A. |
| Author: | Krishnarao dara |
| Affiliation: | thermo Scientific, CoE Lab, Ahmedabad, India |