Find methods for your needs
Application Report: Determination of Bromate, Chloride, and Bromide in Scrubber Water by Ion Chromatography with Suppressed Conductivity Detection
Description
The objective of this application is to analyse bromate in scrubber solution by ion chromatography with suppressed conductivity detection on Integrion HPIC system. The sample matrix contains 0.64% NaCl, 0.16%NaBr, 0.35% Na2CO3, and 0.35% SnO2.2H2O, pH 10.5. This report shows that the bromate in this scrubber solution is very well analysed on IonPac AS19-4um, 2x250mm column using a KOH gradient as eluent. The chloride and bromide can be analysed under the same analytical condition with different sample prepare.| Market: | Environmental |
| Keywords: | DBP, Disinfection, disinfection by product, IonPac AS19-4um |
| Matrix: | Scrubber Water |
| Author: | Linda Lin |
| Affiliation: | Thermo Fisher Scientific |
| Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.