Find methods for your needs
Dionex IonPac AS30 2mm Column Performance Test using its QAR Method
Description
Before running any samples, Thermo Scientific recommends that you first confirm the performance of the column by reproducing the lot validation report chromatogram shipped with column. Compare your results with the one reported in the quality assurance report (QAR). This record provides an eWorkflow for executing the QAR method on an Integrion HPIC system. Other ICS systems can be used for this application.| Market: | Environmental |
| Keywords: | EDA Carbamate, IonPac AS30 column |
| Matrix: | Water |
| Author: | Beibei Huang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.