Find methods for your needs
Improved analysis of carbamates using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of carbamates. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 220 nm.| Market: | Chemical |
| Keywords: | Aldicarb, Carbamates, Carbaryl, Carbofuran, HPLC, Hydroxy carbofuran, Hypersil GOLD, Methiocarb, Methomyl, Naphthol, Oxamyl, Propoxur, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.