Find methods for your needs
Related substances analysis of Ganciclovir as per EP 9.0 monograph with adjustment of chromatographic conditions Be the first to rate this application
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of the related substances of ganciclovir. The analysis was performed on a Thermo Scientific BioBasic SCX HPLC column using the method described in the EP 9.0 monograph. The results obtained for resolution between the peaks due to ganciclovir and its impurity E met the criteria stated in the EP.
Market: Pharma
Keywords: HPLC, UV detection, antiviral, ganciclovir
Matrix: N.A.
Author: krishnarao dara
Affiliation: thermo Scientific, CoE Lab, Ahmedabad, India
Uploaded on 9/16/2019.