Find methods for your needs
HPLC charged aerosol detector analysis of polysorbate 80 and insulin in a biopharmaceutical formulation
Description
This application shows a method for HPLC-CAD analysis of polysorbate 80 and insulin in a biopharmaceutical formulation. The characterization and quantification of polysorbates is difficult because these compounds are heterogeneous mixtures with no chromophore. Because polysorbates are used in final formulations, determination of lot-to-lot variability is critical to both the pharmaceutical and biopharmaceutical industries.| Market: | BioPharma |
| Keywords: | Charged aerosol detector, Polysorbate, Tween, CAD, Biopharmaceuticals |
| Matrix: | biopharmaceutical formulation |
| Author: | Thermo Fisher Scientific |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.